Learn More
Grainger Protective Leg Tip SlipOn Rd 23/64 PK10
Catalog No. 502768571
Encompass_Preferred
The 16ZD89 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 1.4 gallons of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It features an all-steel valve construction and discharge hose, and comes equipped with a vehicle bracket for automotive use. The extinguisher weighs approximately 13 lbs when full and has an operating temperature range of -40°F to 120°F. It uses compressed Novec 1230 gas as the expellant.
Catalog No. 50-276-8571 Supplier Grainger Supplier No. 16ZD89
The 16ZD89 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 1.4 gallons of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It features an all-steel valve construction and discharge hose, and comes equipped with a vehicle bracket for automotive use. The extinguisher weighs approximately 13 lbs when full and has an operating temperature range of -40°F to 120°F. It uses compressed Novec 1230 gas as the expellant.