Learn More
Grainger Protective Leg Tip SlipOn Rd 31/64 PK25
Catalog No. 502765825
Encompass_Preferred
The Grainger 16ZM73 is a Westward brand 1/4-inch drive socket set. It contains 14 SAE sockets ranging from 1/4 inch to 9/16 inch and 14 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a knurled design for oil and slip resistance. This 108-piece set comes organized in a molded case for convenient storage and portability. The sockets have a matte finish and meet or exceed ANSI standards. This is a quality socket set for automotive and light-duty repair applications.
Catalog No. 50-276-5825 Supplier Grainger Supplier No. 16ZM73
The Grainger 16ZM73 is a Westward brand 1/4-inch drive socket set. It contains 14 SAE sockets ranging from 1/4 inch to 9/16 inch and 14 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a knurled design for oil and slip resistance. This 108-piece set comes organized in a molded case for convenient storage and portability. The sockets have a matte finish and meet or exceed ANSI standards. This is a quality socket set for automotive and light-duty repair applications.