Learn More
Grainger Protective Leg Tip SlipOn Rd 47/64 PK25
Catalog No. 502765467
Encompass_Preferred
The 16ZM86 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-A:40-B:C and contains 5 lbs of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It uses a non-ozone depleting agent and leaves no residue, making it ideal for protecting valuable equipment and electronics. The extinguisher has a steel cylinder with epoxy ester lining and is pressurized to 195 psi. It features a chrome-plated brass valve and discharge hose. The extinguisher measures 20.5" high x 5.5" diameter and weighs 16 lbs when full. It has a discharge range of 10-15 ft.
Catalog No. 50-276-5467 Supplier Grainger Supplier No. 16ZM86
The 16ZM86 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-A:40-B:C and contains 5 lbs of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It uses a non-ozone depleting agent and leaves no residue, making it ideal for protecting valuable equipment and electronics. The extinguisher has a steel cylinder with epoxy ester lining and is pressurized to 195 psi. It features a chrome-plated brass valve and discharge hose. The extinguisher measures 20.5" high x 5.5" diameter and weighs 16 lbs when full. It has a discharge range of 10-15 ft.