Learn More
Grainger Protective Leg Tip SlipOn Rd 55/64 PK25
Catalog No. 502765462
Encompass_Preferred
The Grainger 16ZE24 is a Westinghouse 15 Amp, 125 Volt, 3-Wire Grounding Plug with Straight Blades and Side Wire Holes. It has side wire holes that allow for easy installation and removal. This plug features a black thermoplastic housing that offers durability. It can be used on a variety of 15 amp electrical tools and appliances. The plug has UL and CSA certifications for safety and meets OSHA standards. This is a standard 3-prong grounded plug ideal for use on equipment that requires a ground connection.
Catalog No. 50-276-5462 Supplier Grainger Supplier No. 16ZE24
The Grainger 16ZE24 is a Westinghouse 15 Amp, 125 Volt, 3-Wire Grounding Plug with Straight Blades and Side Wire Holes. It has side wire holes that allow for easy installation and removal. This plug features a black thermoplastic housing that offers durability. It can be used on a variety of 15 amp electrical tools and appliances. The plug has UL and CSA certifications for safety and meets OSHA standards. This is a standard 3-prong grounded plug ideal for use on equipment that requires a ground connection.