Learn More
Grainger Protective Leg Tip SlipOn Rd 55/64 PK25
Catalog No. 502765310
Encompass_Preferred
The Grainger 16ZM93 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding. It has a large viewing area for increased visibility. The helmet provides shade levels from 5 to 13 adjustable via external knob. It has adjustable headgear for comfort and optimal fit. The helmet meets ANSI Z87.1 safety standards. The lithium battery provides up to 2000 hours of use. The Speedglas helmet weighs 1.8 lbs and has a 2 year warranty from 3M.
Catalog No. 50-276-5310 Supplier Grainger Supplier No. 16ZM93
The Grainger 16ZM93 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding. It has a large viewing area for increased visibility. The helmet provides shade levels from 5 to 13 adjustable via external knob. It has adjustable headgear for comfort and optimal fit. The helmet meets ANSI Z87.1 safety standards. The lithium battery provides up to 2000 hours of use. The Speedglas helmet weighs 1.8 lbs and has a 2 year warranty from 3M.