Learn More
Cole-Parmer Poly-Net 0437-CRYSTAL Close-mesh high-density polyethylene protective netting, 8" to 10" dia
Catalog No. NC1646013 Shop All Cole-Parmer Products
Encompass_Preferred
This tough, high-density polyethylene (HDPE) resilient protective netting fits snugly over laboratory glassware to absorb impact, reduce breakage, and safeguard against flying glass. Simply cut to desired length to fit most vessels. Use smaller diameter sizes for instruments such as thermometers, pipettes, electrodes, tubes, and syringes. Use larger diameter sizes for bottles, flasks, beakers, funnels, and columns.
Catalog No. NC1646013 Supplier Cole-Parmer Supplier No. 0940545
This tough, high-density polyethylene (HDPE) resilient protective netting fits snugly over laboratory glassware to absorb impact, reduce breakage, and safeguard against flying glass. Simply cut to desired length to fit most vessels. Use smaller diameter sizes for instruments such as thermometers, pipettes, electrodes, tubes, and syringes. Use larger diameter sizes for bottles, flasks, beakers, funnels, and columns.