Learn More
Electron Microscopy Sciences CryoJane Protective Oil, 4oz 4 OZ
SDP
Catalog No. 5030401
Encompass
Used for coating frozen section blocks to prevent dehydration during long-term storage. For use with the CryoJane Tape-Transfer System.
Catalog No. 50-304-01 Supplier Electron Microscopy Sciences Supplier No. 6280074
Used for coating frozen section blocks to prevent dehydration during long-term storage. For use with the CryoJane Tape-Transfer System.