Learn More
HEPATOCHEM Protective Screen for photochemistry
Catalog No. NC1951226
Encompass_Preferred
This plexiglass screen is designed to fit inside standard chemistry hoods and limit direct exposure to photochemical light. The specific color of the screen is 100% effective at blocking all ultraviolet wavelengths. It cuts off light at or below 540 nm wavelength and works great for UV curing operations
Catalog No. NC1951226 Supplier HEPATOCHEM Supplier No. HCK101501002
This plexiglass screen is designed to fit inside standard chemistry hoods and limit direct exposure to photochemical light. The specific color of the screen is 100% effective at blocking all ultraviolet wavelengths. It cuts off light at or below 540 nm wavelength and works great for UV curing operations