Learn More
Grainger Protective Sleeve 25 L 1.34 W Black
Catalog No. 502764042
Encompass_Preferred
The Aeroquip Protective Sleeve Nylon 16P894 is a nylon protective sleeve for hoses and tubes. It is made of abrasion-resistant nylon and has a temperature range of -65°F to +250°F. The sleeve has a 2" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 100'. It protects against cuts, abrasions, and wear and is commonly used on hydraulic lines, air lines, cables, and hoses. The sleeve's nylon construction makes it lightweight, flexible, and durable. It can be cut to desired lengths and has a smooth interior for easy installation over tubing and hose assemblies. The sleeve helps prevent leaks, premature failures, and downtime.
Catalog No. 50-276-4042 Supplier Grainger Supplier No. 16P894
The Aeroquip Protective Sleeve Nylon 16P894 is a nylon protective sleeve for hoses and tubes. It is made of abrasion-resistant nylon and has a temperature range of -65°F to +250°F. The sleeve has a 2" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 100'. It protects against cuts, abrasions, and wear and is commonly used on hydraulic lines, air lines, cables, and hoses. The sleeve's nylon construction makes it lightweight, flexible, and durable. It can be cut to desired lengths and has a smooth interior for easy installation over tubing and hose assemblies. The sleeve helps prevent leaks, premature failures, and downtime.