Learn More
Grainger Protective Sleeve 25 L 1.75 W Black
Catalog No. 502764689
Encompass_Preferred
The Aeroquip Protective Sleeve Nylon 16P895 is a protective sleeve made of nylon that protects hoses, wires and cables from abrasion. It has an operating temperature range of -65°F to +250°F and comes in sizes ranging from 1/4 inch to 1 inch inside diameter, with lengths from 25 feet to 250 feet. The sleeve has good flexibility and kink resistance. It can be used in applications like protecting hydraulic hoses, air hoses, cables and wiring in industrial environments. The nylon material provides durability and protection.
Catalog No. 50-276-4689 Supplier Grainger Supplier No. 16P895
The Aeroquip Protective Sleeve Nylon 16P895 is a protective sleeve made of nylon that protects hoses, wires and cables from abrasion. It has an operating temperature range of -65°F to +250°F and comes in sizes ranging from 1/4 inch to 1 inch inside diameter, with lengths from 25 feet to 250 feet. The sleeve has good flexibility and kink resistance. It can be used in applications like protecting hydraulic hoses, air hoses, cables and wiring in industrial environments. The nylon material provides durability and protection.