Learn More
Grainger Protective Sleeve 25 L 2.63 W Black
Catalog No. 502764140
Encompass_Preferred
The Aeroquip Protective Sleeve Nylon 16P897 is a nylon protective sleeve for hoses and tubes. It is made of abrasion-resistant nylon and has a temperature range of -65°F to +250°F. The sleeve has a 2" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 100'. It protects hoses and tubes from external damage while allowing flexibility. The sleeve's nylon construction makes it lightweight, durable, and resistant to oils, fuels, and solvents. It can be used to protect hydraulic lines, pneumatic lines, and other industrial hoses and tubing. The sleeve is black in color.
Catalog No. 50-276-4140 Supplier Grainger Supplier No. 16P897
The Aeroquip Protective Sleeve Nylon 16P897 is a nylon protective sleeve for hoses and tubes. It is made of abrasion-resistant nylon and has a temperature range of -65°F to +250°F. The sleeve has a 2" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 100'. It protects hoses and tubes from external damage while allowing flexibility. The sleeve's nylon construction makes it lightweight, durable, and resistant to oils, fuels, and solvents. It can be used to protect hydraulic lines, pneumatic lines, and other industrial hoses and tubing. The sleeve is black in color.