Learn More
Grainger Protective Sleeve 25 L 3.38 W Black
Catalog No. 502764690
Encompass_Preferred
The Grainger Protective Sleeve is a 25 foot long, 3.38 inch wide black sleeve made of polyethylene material. It has a maximum temperature rating of 194 degrees Fahrenheit and is designed to provide abrasion resistance and protection for wires, cables, and hoses in indoor and outdoor applications. The sleeve has a smooth inner wall and is sold individually. It can be cut to desired length.
Catalog No. 50-276-4690 Supplier Grainger Supplier No. 16P899
The Grainger Protective Sleeve is a 25 foot long, 3.38 inch wide black sleeve made of polyethylene material. It has a maximum temperature rating of 194 degrees Fahrenheit and is designed to provide abrasion resistance and protection for wires, cables, and hoses in indoor and outdoor applications. The sleeve has a smooth inner wall and is sold individually. It can be cut to desired length.