Learn More
Grainger Protective Sleeve 25 L 4.63 W Black
Catalog No. 502764143
Encompass_Preferred
The Aeroquip Protective Sleeve Nylon 16P901 is a nylon protective sleeve for hoses and tubes. It is made of abrasion-resistant nylon and has a temperature range of -65°F to +250°F. The sleeve has a 2" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 100'. It protects against cuts, abrasions, and wear and is commonly used on hydraulic hoses, air hoses, cables, and more. The sleeve's nylon construction makes it lightweight, flexible, and durable. It can be cut to desired lengths.
Catalog No. 50-276-4143 Supplier Grainger Supplier No. 16P901
The Aeroquip Protective Sleeve Nylon 16P901 is a nylon protective sleeve for hoses and tubes. It is made of abrasion-resistant nylon and has a temperature range of -65°F to +250°F. The sleeve has a 2" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 100'. It protects against cuts, abrasions, and wear and is commonly used on hydraulic hoses, air hoses, cables, and more. The sleeve's nylon construction makes it lightweight, flexible, and durable. It can be cut to desired lengths.