Learn More
Grainger Protective Sleeve 25 L 5.31 W Black
Catalog No. 502764040
Encompass_Preferred
The Aeroquip Protective Sleeve is a nylon sleeve designed to protect hydraulic hoses and fittings from abrasion. It has a maximum temperature rating of 250°F and is resistant to oils, greases, acids, and alkalis. The sleeve's inner diameter measures 1" with a length of 36". It can be cut to desired length. This abrasion sleeve provides excellent protection for hydraulic hoses and fittings in industrial environments.
Catalog No. 50-276-4040 Supplier Grainger Supplier No. 16P902
The Aeroquip Protective Sleeve is a nylon sleeve designed to protect hydraulic hoses and fittings from abrasion. It has a maximum temperature rating of 250°F and is resistant to oils, greases, acids, and alkalis. The sleeve's inner diameter measures 1" with a length of 36". It can be cut to desired length. This abrasion sleeve provides excellent protection for hydraulic hoses and fittings in industrial environments.