Learn More
Med Vet International Instrument Protector 3M Comply 12-1/2 Cm, 13911, 1000/Case
Catalog No. 502829674
Encompass_Preferred
3M™ Comply™ instrument protectors secure delicate instruments during steam and EO sterilization They have 2-mil clear film pockets that prevent the tips from penetrating the pouch Their anti-lock flaps allow proper sterilization of jawed instruments The paper holders allow viewing and identification of surgical instruments Steam and EO indicators verify sterilization is complete They're latex-free A box holds 100 instrument protectors
Catalog No. 50-282-9674 Supplier Med Vet International Supplier No. 155537CS
3M™ Comply™ instrument protectors secure delicate instruments during steam and EO sterilization They have 2-mil clear film pockets that prevent the tips from penetrating the pouch Their anti-lock flaps allow proper sterilization of jawed instruments The paper holders allow viewing and identification of surgical instruments Steam and EO indicators verify sterilization is complete They're latex-free A box holds 100 instrument protectors