Learn More
Sigma Aldrich Fine Chemicals Biosciences Protector RNase Inhibitor
Catalog No. 502906514
Encompass
Protector RNase Inhibitor inactivates a wide spectrum of RNases includingRNase ARNase BRNase T2Thus Protector RNase Inhibitor can help prevent RNase degradation in any application where RNases could cause problems. for instance it canprotect mRNA during cDNA synthesis reactions RT-PCR (in conventional thermal cyclers and qPCR systems) or in vitro transcription/translation reactionsprotect viral RNA during in vitro virus replicationinhibit RNases during RNA isolation and purificationbe used in RNase protection assayshelp prepare RNase-free antibodiesNote Protector RNase Inhibitor does not interfere with enzymes commonly used to prepare or analyze RNA.
Catalog No. 50-290-6514 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3335402001
Protector RNase Inhibitor inactivates a wide spectrum of RNases includingRNase ARNase BRNase T2Thus Protector RNase Inhibitor can help prevent RNase degradation in any application where RNases could cause problems. for instance it canprotect mRNA during cDNA synthesis reactions RT-PCR (in conventional thermal cyclers and qPCR systems) or in vitro transcription/translation reactionsprotect viral RNA during in vitro virus replicationinhibit RNases during RNA isolation and purificationbe used in RNase protection assayshelp prepare RNase-free antibodiesNote Protector RNase Inhibitor does not interfere with enzymes commonly used to prepare or analyze RNA.