Learn More
Sigma Aldrich Fine Chemicals Biosciences Protector RNase Inhibitor | 2000 units
Catalog No. NC1877809
Encompass
Protector RNase Inhibitor (40 U/μl) in storage buffer: 20 mM HEPES-KOH, 50 mM KCl, 8 mM dithiothreitol, 50% glycerol (v/v), pH approximately 7.6 (+4°C).
Catalog No. NC1877809 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3335399001
Protector RNase Inhibitor (40 U/μl) in storage buffer: 20 mM HEPES-KOH, 50 mM KCl, 8 mM dithiothreitol, 50% glycerol (v/v), pH approximately 7.6 (+4°C).