Learn More
Med Vet International Instrument Protector 3M Comply 6-2/3 inch Length, 13913, 100/Pack
Catalog No. 502830506
Encompass_Preferred
Can be used for both steam and EO sterilization Holds the instrument in an open position for proper sterilization For use inside a single peel pouch
Catalog No. 50-283-0506 Supplier Med Vet International Supplier No. 1112461PK
Can be used for both steam and EO sterilization Holds the instrument in an open position for proper sterilization For use inside a single peel pouch