Learn More
FUJIFILM Biosciences ProteFREE ANIMAL 91139 100ML
Catalog No. 502892212
Encompass_Preferred
Enables cell preservation for short-term at -80°C and long-term storage in liquid nitrogen to -196°CHigh post-thaw viability and growthMaintains cell surface marker expression of mesenchymal stem/ stromal cells (MSCs) and PBMCs (T cells) and hematopoietic stem cells (HSCs) post-thawComplete, ready-to-use mediumContains DMSO in the formulationBy purchasing this product you are agree that you will not use this product as excipient
Catalog No. 50-289-2212 Supplier FUJIFILM Biosciences Supplier No. 91139100ML
Enables cell preservation for short-term at -80°C and long-term storage in liquid nitrogen to -196°CHigh post-thaw viability and growthMaintains cell surface marker expression of mesenchymal stem/ stromal cells (MSCs) and PBMCs (T cells) and hematopoietic stem cells (HSCs) post-thawComplete, ready-to-use mediumContains DMSO in the formulationBy purchasing this product you are agree that you will not use this product as excipient