Learn More
MSC Honeywell Uvex Protege Safety Glasses, Polycarbonate, Amber Lens, Metallic Black Frame, 1 ea
Catalog No. 5000508661
Encompass
These safety glasses are designed for high-impact protection in industrial environments, featuring a lightweight, half-frame wraparound design with an amber lens for enhanced visibility. Engineered for durability and comfort, the polycarbonate construction offers superior optical clarity and reliable defense against hazardous debris.
  • HydroShield® anti-fog coating provides consistent performance that lasts 90X longer than standard anti-fog products
  • Delivers 2X greater scratch resistance compared to standard anti-fog coatings
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards
  • Passes military ballistic impact tests
  • Offers 99.9% UVA and UVB protection
  • Features a universal fit with soft, flexible nose fingers and soft temple pads for extended wear comfort
  • Includes a lifetime frame guarantee
Catalog No. 50-005-08661 Supplier MSC Supplier No. 32474637
These safety glasses are designed for high-impact protection in industrial environments, featuring a lightweight, half-frame wraparound design with an amber lens for enhanced visibility. Engineered for durability and comfort, the polycarbonate construction offers superior optical clarity and reliable defense against hazardous debris.
  • HydroShield® anti-fog coating provides consistent performance that lasts 90X longer than standard anti-fog products
  • Delivers 2X greater scratch resistance compared to standard anti-fog coatings
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards
  • Passes military ballistic impact tests
  • Offers 99.9% UVA and UVB protection
  • Features a universal fit with soft, flexible nose fingers and soft temple pads for extended wear comfort
  • Includes a lifetime frame guarantee