Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01&B2M&AFP (FMNKFIYEI) Complex Protein (Monomer, MALS verified)
Catalog No. 502331295
Encompass_Preferred
Biotinylated Human HLA-A*02:01&B2M&AFP (FMNKFIYEI) Complex Protein (HLA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) & FMNKFIYEI peptide (Accession # AAA59606.1 (HLA-A*02:01) & P61769 (B2M) & FMNKFIYEI).
Catalog No. 50-233-1295 Supplier ACROBiosystems Supplier No. HLAH82E325UG
Biotinylated Human HLA-A*02:01&B2M&AFP (FMNKFIYEI) Complex Protein (HLA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) & FMNKFIYEI peptide (Accession # AAA59606.1 (HLA-A*02:01) & P61769 (B2M) & FMNKFIYEI).