Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01 & B2M complex with MAGE-A1 peptide (KVLEYVIKV), monomeric, Avitag biotinylated, 25 μg
Catalog No. 502370198
Encompass_Preferred
Biotinylated human HLA-A*02:01&B2M complex loaded with the MAGE-A1 peptide KVLEYVIKV. Expressed in HEK293 cells and enzymatically biotinylated via Avitag, this monomeric, MALS-verified complex is supplied lyophilized for use in antibody binding and immunoassay workflows.
  • Expressed in HEK293 (human) cells for human-like glycosylation.
  • Monomeric, MALS-verified format for accurate molecular characterization.
  • Enzymatically biotinylated via Avitag for streptavidin capture and immobilization.
  • Supplied lyophilized from PBS with trehalose as protectant for stability.
  • High purity suitable for immunoassays (>90% by SDS-PAGE; >95% by SEC-MALS).
  • Validated in antibody binding assays at 1 μg/mL (demonstrated linear ranges for anti-HLA and anti-B2M antibodies).
  • Stable when stored lyophilized at -20°C to -70°C; avoid repeated freeze-thaw cycles.
Catalog No. 50-237-0198 Supplier ACROBiosystems Supplier No. HLMH82E125UG
Biotinylated human HLA-A*02:01&B2M complex loaded with the MAGE-A1 peptide KVLEYVIKV. Expressed in HEK293 cells and enzymatically biotinylated via Avitag, this monomeric, MALS-verified complex is supplied lyophilized for use in antibody binding and immunoassay workflows.
  • Expressed in HEK293 (human) cells for human-like glycosylation.
  • Monomeric, MALS-verified format for accurate molecular characterization.
  • Enzymatically biotinylated via Avitag for streptavidin capture and immobilization.
  • Supplied lyophilized from PBS with trehalose as protectant for stability.
  • High purity suitable for immunoassays (>90% by SDS-PAGE; >95% by SEC-MALS).
  • Validated in antibody binding assays at 1 μg/mL (demonstrated linear ranges for anti-HLA and anti-B2M antibodies).
  • Stable when stored lyophilized at -20°C to -70°C; avoid repeated freeze-thaw cycles.