Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems PE-Labeled Human HLA-A*02:01 & B2M & NY-ESO-1 (SLLMWITQV) Tetramer Protein, 200 tests (HL1-HP2E6-200TESTS)

Catalog No. 502370207
Encompass_Preferred

Pre-assembled PE-conjugated HLA-A*02:01/B2M tetramer presenting the NY-ESO-1 (SLLMWITQV) peptide for detecting antigen-specific T cells and evaluating binding activity in flow cytometry and immunoassays. Supplied lyophilized for long-term storage; reconstitute following the Certificate of Analysis.

  • Conjugate: PE.
  • Intended use: detection of NY-ESO-1-specific T cells and binding assays.
  • Format: pre-assembled tetramer assembled from biotinylated monomers and PE-streptavidin.
  • Expression system: human HEK293 cells.
  • Storage: lyophilized at -20°C to -70°C; protect from light and avoid repeated freeze-thaw cycles.
  • Buffer: lyophilized from 0.22 μm filtered solution in 0.2% BSA with trehalose.
  • Recommended assay conditions: ELISA immobilized at 1 μg/mL; FACS 1:25 dilution.
  • Stability: 12 months lyophilized; up to 3 months after reconstitution when stored at -70°C.

Catalog No. 50-237-0207 Supplier ACROBiosystems Supplier No. HL1HP2E6200TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pre-assembled PE-conjugated HLA-A*02:01/B2M tetramer presenting the NY-ESO-1 (SLLMWITQV) peptide for detecting antigen-specific T cells and evaluating binding activity in flow cytometry and immunoassays. Supplied lyophilized for long-term storage; reconstitute following the Certificate of Analysis.

  • Conjugate: PE.
  • Intended use: detection of NY-ESO-1-specific T cells and binding assays.
  • Format: pre-assembled tetramer assembled from biotinylated monomers and PE-streptavidin.
  • Expression system: human HEK293 cells.
  • Storage: lyophilized at -20°C to -70°C; protect from light and avoid repeated freeze-thaw cycles.
  • Buffer: lyophilized from 0.22 μm filtered solution in 0.2% BSA with trehalose.
  • Recommended assay conditions: ELISA immobilized at 1 μg/mL; FACS 1:25 dilution.
  • Stability: 12 months lyophilized; up to 3 months after reconstitution when stored at -70°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.