Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01 & B2M Complex With MAGE-A4 Peptide (GVYDGREHTV), Monomer, 200 μg
Catalog No. 502370222
Encompass_Preferred
Recombinant biotinylated human HLA-A*02:01 complexed with beta-2-microglobulin and the MAGE-A4 peptide (GVYDGREHTV). Supplied as a monomer and verified by SEC-MALS, the product is intended for binding assays, antibody characterization, and T-cell epitope research.
  • Biotinylated via site-specific Avitag enzymatic labeling for consistent orientation in binding assays.
  • Monomeric and SEC-MALS verified to confirm proper assembly.
  • Lyophilized presentation with trehalose for enhanced stability during storage.
  • Stable long-term when stored frozen; minimizes freeze-thaw cycles after reconstitution.
  • Suitable for ELISA, SPR/Biacore, and antibody or T-cell epitope studies targeting MAGE-A4.
Catalog No. 50-237-0222 Supplier ACROBiosystems Supplier No. HLMH82E5200UG
Recombinant biotinylated human HLA-A*02:01 complexed with beta-2-microglobulin and the MAGE-A4 peptide (GVYDGREHTV). Supplied as a monomer and verified by SEC-MALS, the product is intended for binding assays, antibody characterization, and T-cell epitope research.
  • Biotinylated via site-specific Avitag enzymatic labeling for consistent orientation in binding assays.
  • Monomeric and SEC-MALS verified to confirm proper assembly.
  • Lyophilized presentation with trehalose for enhanced stability during storage.
  • Stable long-term when stored frozen; minimizes freeze-thaw cycles after reconstitution.
  • Suitable for ELISA, SPR/Biacore, and antibody or T-cell epitope studies targeting MAGE-A4.