Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Alexa Fluor 488-Labeled Human HLA-A*11:01 & B2M & KRASG12D (VVGADGVGK) Complex Protein (Monomer), 200 μg

Catalog No. 502370230
Encompass_Preferred

Alexa Fluor 488-labeled human HLA-A*11:01 complexed with beta-2 microglobulin and the KRAS G12D peptide (VVGADGVGK). Monomeric complex produced in HEK293 cells and conjugated with AF488 for fluorescence-based assays such as flow cytometry and staining. Lyophilized formulation with trehalose for improved stability.

  • Alexa Fluor 488 conjugate; excitation 488 nm, emission 517 nm
  • Monomeric HLA-A*11:01 complexed with B2M and KRAS G12D peptide (VVGADGVGK)
  • Expression system: HEK293 (human 293) cells
  • Lyophilized from PBS, pH 7.4, with trehalose
  • Size: 200 μg
  • Purity greater than 90% by SDS-PAGE
  • Contains HLA-A*11:01 (Gly25-Thr305) and B2M (Ile21-Met119) with C-terminal His tag
  • Store lyophilized at -20°C or lower; protect from light and avoid repeated freeze-thaw cycles

Catalog No. 50-237-0230 Supplier ACROBiosystems Supplier No. HLDHA2H7200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor 488-labeled human HLA-A*11:01 complexed with beta-2 microglobulin and the KRAS G12D peptide (VVGADGVGK). Monomeric complex produced in HEK293 cells and conjugated with AF488 for fluorescence-based assays such as flow cytometry and staining. Lyophilized formulation with trehalose for improved stability.

  • Alexa Fluor 488 conjugate; excitation 488 nm, emission 517 nm
  • Monomeric HLA-A*11:01 complexed with B2M and KRAS G12D peptide (VVGADGVGK)
  • Expression system: HEK293 (human 293) cells
  • Lyophilized from PBS, pH 7.4, with trehalose
  • Size: 200 μg
  • Purity greater than 90% by SDS-PAGE
  • Contains HLA-A*11:01 (Gly25-Thr305) and B2M (Ile21-Met119) with C-terminal His tag
  • Store lyophilized at -20°C or lower; protect from light and avoid repeated freeze-thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.