Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human HLA-A*11:01 & β2-microglobulin Complex Protein (KRAS VVGAGGVGK), 200 μg

Catalog No. 502370233
Encompass_Preferred

Biotinylated human HLA-A*11:01-β2-microglobulin complex loaded with the KRAS peptide VVGAGGVGK. The monomeric, MALS-verified protein is expressed in HEK293 cells and supplied lyophilized in 200 μg (100 μg x 2) vials for use in ELISA and other binding assays.

  • Biotinylated with C-terminal His tag and Avi tag (Avitag).
  • Expression system: Human 293 (HEK293) cells.
  • Unit size: 200 μg (100 μg x 2).
  • Purity greater than 90% by SDS-PAGE and SEC-MALS.
  • Observed SDS-PAGE migration: 40-45 kDa and 12 kDa under reducing conditions due to glycosylation.
  • Formulation: Lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose as protectant.
  • Storage: Lyophilized at -20°C to -70°C (stable 12 months); reconstituted storage at -70°C (stable 3 months under sterile conditions).

Catalog No. 50-237-0233 Supplier ACROBiosystems Supplier No. HLSH82E3200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated human HLA-A*11:01-β2-microglobulin complex loaded with the KRAS peptide VVGAGGVGK. The monomeric, MALS-verified protein is expressed in HEK293 cells and supplied lyophilized in 200 μg (100 μg x 2) vials for use in ELISA and other binding assays.

  • Biotinylated with C-terminal His tag and Avi tag (Avitag).
  • Expression system: Human 293 (HEK293) cells.
  • Unit size: 200 μg (100 μg x 2).
  • Purity greater than 90% by SDS-PAGE and SEC-MALS.
  • Observed SDS-PAGE migration: 40-45 kDa and 12 kDa under reducing conditions due to glycosylation.
  • Formulation: Lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose as protectant.
  • Storage: Lyophilized at -20°C to -70°C (stable 12 months); reconstituted storage at -70°C (stable 3 months under sterile conditions).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.