Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human HLA-A*02:01 & B2M Complex Protein with MAGE-A3 (KVAELVHFL), Monomer, MALS Verified, His-Avi Tagged, 25 μg

Catalog No. 502370245
Encompass_Preferred

Biotinylated human HLA-A*02:01 complexed with beta-2 microglobulin and the MAGE-A3 peptide (KVAELVHFL). The monomeric complex is enzymatically biotinylated at an AviTag, produced in HEK293 cells, lyophilized for stability, and characterized by SEC-MALS and SDS-PAGE for use in binding assays, immunology studies, and reagent development.

  • Contains HLA-A*02:01 (Gly25-Ile308), B2M (Ile21-Met119), and MAGE-A3 peptide KVAELVHFL.
  • Enzymatically biotinylated at the AviTag lysine; carries a C-terminal His tag and Avi tag.
  • Produced in HEK293 cells and lyophilized from PBS with trehalose for preservation.
  • Purity >90% by SDS-PAGE; >95% by SEC-MALS.
  • Endotoxin <1.0 EU/μg (LAL), suitable for binding assays.
  • Apparent molecular masses: 38-44 kDa (HLA) and ~12 kDa (B2M) under reducing SDS-PAGE.
  • Storage: stable lyophilized at -20°C to -70°C for 12 months; reconstituted storage at -70°C for 3 months.

Catalog No. 50-237-0245 Supplier ACROBiosystems Supplier No. HLMH82E325UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated human HLA-A*02:01 complexed with beta-2 microglobulin and the MAGE-A3 peptide (KVAELVHFL). The monomeric complex is enzymatically biotinylated at an AviTag, produced in HEK293 cells, lyophilized for stability, and characterized by SEC-MALS and SDS-PAGE for use in binding assays, immunology studies, and reagent development.

  • Contains HLA-A*02:01 (Gly25-Ile308), B2M (Ile21-Met119), and MAGE-A3 peptide KVAELVHFL.
  • Enzymatically biotinylated at the AviTag lysine; carries a C-terminal His tag and Avi tag.
  • Produced in HEK293 cells and lyophilized from PBS with trehalose for preservation.
  • Purity >90% by SDS-PAGE; >95% by SEC-MALS.
  • Endotoxin <1.0 EU/μg (LAL), suitable for binding assays.
  • Apparent molecular masses: 38-44 kDa (HLA) and ~12 kDa (B2M) under reducing SDS-PAGE.
  • Storage: stable lyophilized at -20°C to -70°C for 12 months; reconstituted storage at -70°C for 3 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.