Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human HLA-A*11:01 & B2M & KRASG12D (VVGADGVGK) Complex Protein, His tag, 200 μg

Catalog No. 502370201
Encompass_Preferred

Human HLA-A*11:01 & B2M & KRASG12D (VVGADGVGK) Complex Protein is a monomeric MHC class I complex composed of the HLA-A*11:01 heavy chain, beta-2-microglobulin, and the KRAS G12D-derived peptide. It is expressed in HEK293 cells, carries a C-terminal His tag, and is provided lyophilized for biophysical and bioactivity applications.

  • Expressed in human 293 cells (HEK293).
  • Contains residues Gly 25-Thr 305 (HLA-A*11:01) and Ile 21-Met 119 (B2M), plus peptide VVGADGVGK.
  • C-terminal polyhistidine tag for purification or detection.
  • Purity greater than 90% as determined by SDS-PAGE and SEC-MALS.
  • MALS-verified molecular weight and monomeric state.
  • Lyophilized formulation in PBS with trehalose; 0.22 μm filtered prior to lyophilization.
  • Store lyophilized at -20°C or lower; stable up to 12 months lyophilized.
  • Endotoxin less than 1.0 EU/μg (LAL or rFC).
  • Suitable for ELISA, SPR, and antibody binding assays.

Catalog No. 50-237-0201 Supplier ACROBiosystems Supplier No. HLDH52H4200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human HLA-A*11:01 & B2M & KRASG12D (VVGADGVGK) Complex Protein is a monomeric MHC class I complex composed of the HLA-A*11:01 heavy chain, beta-2-microglobulin, and the KRAS G12D-derived peptide. It is expressed in HEK293 cells, carries a C-terminal His tag, and is provided lyophilized for biophysical and bioactivity applications.

  • Expressed in human 293 cells (HEK293).
  • Contains residues Gly 25-Thr 305 (HLA-A*11:01) and Ile 21-Met 119 (B2M), plus peptide VVGADGVGK.
  • C-terminal polyhistidine tag for purification or detection.
  • Purity greater than 90% as determined by SDS-PAGE and SEC-MALS.
  • MALS-verified molecular weight and monomeric state.
  • Lyophilized formulation in PBS with trehalose; 0.22 μm filtered prior to lyophilization.
  • Store lyophilized at -20°C or lower; stable up to 12 months lyophilized.
  • Endotoxin less than 1.0 EU/μg (LAL or rFC).
  • Suitable for ELISA, SPR, and antibody binding assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.