Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human HLA-A*02:01 & B2M & RHAMM-R3 (ILSLELMKL) Complex Protein (Monomer, MALS verified)

Catalog No. 502370203
Encompass_Preferred

Biotinylated Human HLA-A*02:01 & B2M & RHAMM-R3 (ILSLELMKL) Complex Protein is a monomeric MHC class I complex produced in human HEK293 cells and loaded with the RHAMM-R3 peptide. The product is site-specifically biotinylated, MALS-verified as monomeric, and supplied as a 25 μg solution formulated in PBS with trehalose for use in T-cell assays, immunology research, and binding studies.

  • Monomer, MALS verified.
  • Site-specific enzymatic biotinylation.
  • Purity greater than 90% by SEC-MALS and SDS-PAGE.
  • Contains RHAMM-R3 peptide (ILSLELMKL).
  • Supplied as 25 μg in 0.2 μm filtered PBS, pH 7.4, with trehalose protectant.
  • Store at -70°C or lower; avoid repeated freeze-thaw cycles.

Catalog No. 50-237-0203 Supplier ACROBiosystems Supplier No. HLRH82E625UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated Human HLA-A*02:01 & B2M & RHAMM-R3 (ILSLELMKL) Complex Protein is a monomeric MHC class I complex produced in human HEK293 cells and loaded with the RHAMM-R3 peptide. The product is site-specifically biotinylated, MALS-verified as monomeric, and supplied as a 25 μg solution formulated in PBS with trehalose for use in T-cell assays, immunology research, and binding studies.

  • Monomer, MALS verified.
  • Site-specific enzymatic biotinylation.
  • Purity greater than 90% by SEC-MALS and SDS-PAGE.
  • Contains RHAMM-R3 peptide (ILSLELMKL).
  • Supplied as 25 μg in 0.2 μm filtered PBS, pH 7.4, with trehalose protectant.
  • Store at -70°C or lower; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.