Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human HLA-A*11:01 & B2M & KRAS (VVGAGGVGK) Complex Protein (Monomer, MALS verified), 200 μg

Catalog No. 502370205
Encompass_Preferred

Recombinant human HLA-A*11:01 complexed with beta-2 microglobulin (B2M) and the KRAS peptide VVGAGGVGK. This monomeric MHC class I complex is produced in human 293 cells (HEK293) and verified by multi-angle light scattering (MALS). The product is supplied lyophilized with trehalose as a stabilizer and is available in research sizes.

  • Expressed in human 293 cells (HEK293).
  • Contains HLA-A*11:01 (Gly25-Thr305), B2M (Ile21-Met119), and KRAS peptide VVGAGGVGK.
  • Monomeric complex verified by multi-angle light scattering (MALS).
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Available in 25 μg and 200 μg sizes.

Catalog No. 50-237-0205 Supplier ACROBiosystems Supplier No. HLSH52H5200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human HLA-A*11:01 complexed with beta-2 microglobulin (B2M) and the KRAS peptide VVGAGGVGK. This monomeric MHC class I complex is produced in human 293 cells (HEK293) and verified by multi-angle light scattering (MALS). The product is supplied lyophilized with trehalose as a stabilizer and is available in research sizes.

  • Expressed in human 293 cells (HEK293).
  • Contains HLA-A*11:01 (Gly25-Thr305), B2M (Ile21-Met119), and KRAS peptide VVGAGGVGK.
  • Monomeric complex verified by multi-angle light scattering (MALS).
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Available in 25 μg and 200 μg sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.