Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems PE-Labeled Human HLA-A*02:01 & β2-Microglobulin Tetramer Protein (HPV16-E6 TIHDIILECV), PE conjugate, 200 μg

Catalog No. 502370209
Encompass_Preferred

PE-labeled HLA-A*02:01/β2-microglobulin tetramer loaded with the HPV16-E6 peptide (TIHDIILECV). Assembled from biotinylated monomers and PE-labeled streptavidin, the complex is expressed in HEK293 cells and supplied in PBS with 1% BSA and trehalose for stabilization.

  • PE conjugated for fluorescence detection.
  • Contains HLA-A*02:01 complexed with β2-microglobulin and HPV16-E6 peptide.
  • Expressed in HEK293 (human 293) cells.
  • Includes C-terminal His tag and Avi tag (Avitag).
  • Supplied in PBS, 1% BSA, pH 7.4, 0.2 μm filtered, with trehalose protectant.
  • Available unit sizes: 25 μg and 200 μg.
  • Store at -70 °C; protect from light and avoid repeated freeze-thaw cycles.
  • Excitation 488 nm / 561 nm; emission 575 nm.
  • Validated for ELISA/binding studies (anti-B2M linear range 0.1-2 ng/mL).

Catalog No. 50-237-0209 Supplier ACROBiosystems Supplier No. HLHHP2H7200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PE-labeled HLA-A*02:01/β2-microglobulin tetramer loaded with the HPV16-E6 peptide (TIHDIILECV). Assembled from biotinylated monomers and PE-labeled streptavidin, the complex is expressed in HEK293 cells and supplied in PBS with 1% BSA and trehalose for stabilization.

  • PE conjugated for fluorescence detection.
  • Contains HLA-A*02:01 complexed with β2-microglobulin and HPV16-E6 peptide.
  • Expressed in HEK293 (human 293) cells.
  • Includes C-terminal His tag and Avi tag (Avitag).
  • Supplied in PBS, 1% BSA, pH 7.4, 0.2 μm filtered, with trehalose protectant.
  • Available unit sizes: 25 μg and 200 μg.
  • Store at -70 °C; protect from light and avoid repeated freeze-thaw cycles.
  • Excitation 488 nm / 561 nm; emission 575 nm.
  • Validated for ELISA/binding studies (anti-B2M linear range 0.1-2 ng/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.