Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01 and B2M and MAGE-A4 (GVYDGREHTV) Complex Protein, Monomer, Avitag, 25 μg
Supplier: ACROBiosystems HLMH82E525UG
Biotinylated Human HLA-A*02:01 and B2M and MAGE-A4 (GVYDGREHTV) complex is a monomeric MHC class I protein intended for antibody binding assays and assay development. Expressed in HEK293 cells, enzymatically biotinylated for site-specific labeling, and supplied lyophilized with trehalose; validated by SEC-MALS for purity and molecular weight.
- Expressed in human HEK293 cells.
- Contains HLA-A*02:01 (Gly25-Ile308), B2M (Ile21-Met119), and GVYDGREHTV peptide.
- Enzymatic site-specific biotinylation for consistent labeling.
- Monomeric and MALS-verified; >95% purity by SEC-MALS.
- Lyophilized in PBS with trehalose; stable at -20°C to -70°C in lyophilized state.
- Supplied as 25 μg; COA and datasheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.