Learn More
ACROBiosystems PE-Labeled Human HLA-A*02:01 & B2M & NY-ESO-1 (SLLMWITQC) Tetramer Protein, 25 tests
Catalog No. 502370225
Encompass_Preferred
PE-labeled HLA-A*02:01 & B2M & NY-ESO-1 (SLLMWITQC) tetramer protein for detection of NY-ESO-1-specific CD8+ T cells by flow cytometry. Supplied lyophilized as a preassembled tetrameric complex produced in human HEK293 cells; the complex carries a C-terminal His tag and an Avi tag. Certificate of Analysis provides reconstitution and storage instructions.
  • Preassembled PE-labeled tetrameric complex for direct staining
  • Optimized for detection of NY-ESO-1-specific CD8+ T cells by flow cytometry
  • Lyophilized formulation for extended shelf life
  • Contains C-terminal His tag and Avi tag for quality control
  • Supplied for 25 tests; refer to the Certificate of Analysis for reconstitution
  • Store lyophilized at -20°C to -70°C and protect from light
Catalog No. 50-237-0225 Supplier ACROBiosystems Supplier No. HL1HP2E525TESTS
PE-labeled HLA-A*02:01 & B2M & NY-ESO-1 (SLLMWITQC) tetramer protein for detection of NY-ESO-1-specific CD8+ T cells by flow cytometry. Supplied lyophilized as a preassembled tetrameric complex produced in human HEK293 cells; the complex carries a C-terminal His tag and an Avi tag. Certificate of Analysis provides reconstitution and storage instructions.
  • Preassembled PE-labeled tetrameric complex for direct staining
  • Optimized for detection of NY-ESO-1-specific CD8+ T cells by flow cytometry
  • Lyophilized formulation for extended shelf life
  • Contains C-terminal His tag and Avi tag for quality control
  • Supplied for 25 tests; refer to the Certificate of Analysis for reconstitution
  • Store lyophilized at -20°C to -70°C and protect from light