Learn More
ACROBiosystems Human HLA-A*11:01 & B2M Tetramer presenting KRAS G12V (VVGAVGVGK), 200 μg
Catalog No. 502370234
Encompass_Preferred
Tetrameric HLA-A*11:01 and beta-2 microglobulin complex presenting the KRAS G12V peptide (VVGAVGVGK), supplied lyophilized as 200 μg for immunoassays and reagent development.
  • Expressed in human HEK293 cells.
  • Includes HLA-A*11:01 (Gly25-Thr305), B2M (Ile21-Met119), and KRAS G12V peptide VVGAVGVGK.
  • Assembled as a biotinylated tetramer via streptavidin.
  • Features C-terminal His tag and Avi tag for purification and biotinylation.
  • Lyophilized from PBS, pH 7.4, with trehalose protectant.
  • Purity greater than 90% by SDS-PAGE; endotoxin <1.0 EU/μg.
  • Stable 12 months lyophilized at -20°C to -70°C; 3 months at -70°C after reconstitution.
  • Bioactivity validated by ELISA for antibody binding.
Catalog No. 50-237-0234 Supplier ACROBiosystems Supplier No. HLVH52H8200UG
Tetrameric HLA-A*11:01 and beta-2 microglobulin complex presenting the KRAS G12V peptide (VVGAVGVGK), supplied lyophilized as 200 μg for immunoassays and reagent development.
  • Expressed in human HEK293 cells.
  • Includes HLA-A*11:01 (Gly25-Thr305), B2M (Ile21-Met119), and KRAS G12V peptide VVGAVGVGK.
  • Assembled as a biotinylated tetramer via streptavidin.
  • Features C-terminal His tag and Avi tag for purification and biotinylation.
  • Lyophilized from PBS, pH 7.4, with trehalose protectant.
  • Purity greater than 90% by SDS-PAGE; endotoxin <1.0 EU/μg.
  • Stable 12 months lyophilized at -20°C to -70°C; 3 months at -70°C after reconstitution.
  • Bioactivity validated by ELISA for antibody binding.