Learn More
ACROBiosystems Alexa Fluor 488-Labeled Human HLA-A*11:01 & B2M Complex Protein (KRAS G12D VVGADGVGK), Monomer, 25 μg
Supplier: ACROBiosystems HLDHA2H725UG
Alexa Fluor 488-labeled monomeric HLA-A*11:01/B2M complex presenting the KRAS G12D peptide (VVGADGVGK). Recombinantly expressed in HEK293 cells, C-terminal His-tagged, and conjugated on primary amines with Alexa Fluor 488 for fluorescence detection and antibody-binding assays.
- Expression host: HEK293 human cells.
- Conjugate: Alexa Fluor 488 (excitation 488 nm; emission 517 nm).
- Tag: C-terminal polyhistidine.
- Available sizes: 25 μg and 200 μg, lyophilized.
- Formulation: PBS pH 7.4 with trehalose, lyophilized.
- Storage: store lyophilized at -20°C or lower; stable -20°C to -70°C for 12 months.
- Molecular weight: calculated 36.4 kDa and 14.0 kDa; migrates ~55-60 kDa and <14 kDa on reducing SDS-PAGE.
- Recommended use: fluorescence detection and antibody binding assays; ELISA tested at 1 μg/mL (100 μL/well).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.