Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human HLA-A*02:01 & B2M & MAGE-A4 (KVLEHVVRV) Tetramer Protein, 25 μg (HLP-HP2H3-25UG)
Catalog No. 502370199
Encompass_Preferred
PE-labeled HLA-A*02:01-B2M-MAGE-A4 (KVLEHVVRV) tetramer protein is a preformed MHC class I complex designed for direct detection of MAGE-A4-specific CD8+ T cells by flow cytometry. The reagent combines HLA-A*02:01 heavy chain, beta-2 microglobulin, and the KVLEHVVRV peptide, is produced in HEK293 cells, and is supplied lyophilized for convenient storage and assay setup.
  • PE-labeled for direct flow cytometry detection.
  • Preformed tetramer containing HLA-A*02:01, B2M, and the MAGE-A4 peptide KVLEHVVRV.
  • Produced in HEK293 (human 293) expression system.
  • Lyophilized format for stability; contains trehalose protectant.
  • Supplied in PBS with 1% BSA, pH 7.4.
  • Purity >90% as determined by SDS-PAGE.
  • Size: 25 μg, ready for reconstitution.
Catalog No. 50-237-0199 Supplier ACROBiosystems Supplier No. HLPHP2H325UG
PE-labeled HLA-A*02:01-B2M-MAGE-A4 (KVLEHVVRV) tetramer protein is a preformed MHC class I complex designed for direct detection of MAGE-A4-specific CD8+ T cells by flow cytometry. The reagent combines HLA-A*02:01 heavy chain, beta-2 microglobulin, and the KVLEHVVRV peptide, is produced in HEK293 cells, and is supplied lyophilized for convenient storage and assay setup.
  • PE-labeled for direct flow cytometry detection.
  • Preformed tetramer containing HLA-A*02:01, B2M, and the MAGE-A4 peptide KVLEHVVRV.
  • Produced in HEK293 (human 293) expression system.
  • Lyophilized format for stability; contains trehalose protectant.
  • Supplied in PBS with 1% BSA, pH 7.4.
  • Purity >90% as determined by SDS-PAGE.
  • Size: 25 μg, ready for reconstitution.