Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human HLA-A*11:01 & B2M & KRASG12V (VVGAVGVGK) Complex Protein (Monomer, MALS verified) - 200 μg

Catalog No. 502370206
Encompass_Preferred

Recombinant human HLA-A*11:01 and beta-2 microglobulin (B2M) complexed with the KRASG12V peptide (VVGAVGVGK). Produced in HEK293 cells, the product is supplied lyophilized with a C-terminal His tag and verified as a monomer by SEC-MALS for use in antigen presentation and antibody-binding assays.

  • Expression system: HEK293 (human 293) cells.
  • Format: complex protein, monomer verified by SEC-MALS.
  • Purity: greater than 90% by SDS-PAGE and SEC-MALS.
  • Molecular weight: migrates ~40-45 kDa and ~12 kDa by SDS-PAGE; ~45-60 kDa by SEC-MALS.
  • Tag: C-terminal polyhistidine tag.
  • Storage: lyophilized from PBS, pH 7.4 with trehalose; stable 12 months at -20°C to -70°C in lyophilized state.
  • Endotoxin: less than 1.0 EU per μg (LAL/rFC).
  • Applications: suitable for antigen presentation studies and antibody-binding assays; can be immobilized for ELISA binding with high sensitivity.

Catalog No. 50-237-0206 Supplier ACROBiosystems Supplier No. HLVH52H3200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human HLA-A*11:01 and beta-2 microglobulin (B2M) complexed with the KRASG12V peptide (VVGAVGVGK). Produced in HEK293 cells, the product is supplied lyophilized with a C-terminal His tag and verified as a monomer by SEC-MALS for use in antigen presentation and antibody-binding assays.

  • Expression system: HEK293 (human 293) cells.
  • Format: complex protein, monomer verified by SEC-MALS.
  • Purity: greater than 90% by SDS-PAGE and SEC-MALS.
  • Molecular weight: migrates ~40-45 kDa and ~12 kDa by SDS-PAGE; ~45-60 kDa by SEC-MALS.
  • Tag: C-terminal polyhistidine tag.
  • Storage: lyophilized from PBS, pH 7.4 with trehalose; stable 12 months at -20°C to -70°C in lyophilized state.
  • Endotoxin: less than 1.0 EU per μg (LAL/rFC).
  • Applications: suitable for antigen presentation studies and antibody-binding assays; can be immobilized for ELISA binding with high sensitivity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.