Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01-B2M Complex Loaded With MAGE-A2 Peptide (YLQLVFGIEV), Monomer, Biotin/Avi- and His-Tagged, 25 μg
Catalog No. 502370235
Encompass_Preferred
This biotinylated monomeric peptide-MHC complex comprises human HLA-A*02:01 co-expressed with beta-2 microglobulin and loaded with the MAGE-A2 peptide YLQLVFGIEV. Produced in HEK293 cells and verified by multi-angle light scattering (MALS), it is intended for tetramer assembly and antigen-specific T cell detection.
  • Biotinylated with Avi tag for streptavidin conjugation.
  • Contains HLA-A*02:01, beta-2 microglobulin, and MAGE-A2 (YLQLVFGIEV) peptide.
  • Expressed in human HEK293 cells.
  • Verified monomeric status by MALS.
  • Calculated molecular weight approximately 36.3 kDa (HLA) and 11.7 kDa (B2M); migrates ~38-44 kDa and ~12 kDa under reducing conditions.
  • Supplied at 25 μg for research use.
Catalog No. 50-237-0235 Supplier ACROBiosystems Supplier No. HLMH82E625UG
This biotinylated monomeric peptide-MHC complex comprises human HLA-A*02:01 co-expressed with beta-2 microglobulin and loaded with the MAGE-A2 peptide YLQLVFGIEV. Produced in HEK293 cells and verified by multi-angle light scattering (MALS), it is intended for tetramer assembly and antigen-specific T cell detection.
  • Biotinylated with Avi tag for streptavidin conjugation.
  • Contains HLA-A*02:01, beta-2 microglobulin, and MAGE-A2 (YLQLVFGIEV) peptide.
  • Expressed in human HEK293 cells.
  • Verified monomeric status by MALS.
  • Calculated molecular weight approximately 36.3 kDa (HLA) and 11.7 kDa (B2M); migrates ~38-44 kDa and ~12 kDa under reducing conditions.
  • Supplied at 25 μg for research use.