Learn More
ACROBiosystems Biotinylated Human HLA-A*11:01 & B2M Complex with KRAS (VVGAGGVGK), Monomer, 25 μg
Supplier: ACROBiosystems HLSH82E325UG
Biotinylated human HLA-A*11:01 and beta-2 microglobulin (B2M) complex loaded with KRAS peptide VVGAGGVGK. Expressed in HEK293 cells and verified as a monomer by SEC-MALS, this lyophilized 25 μg reagent is suitable for antibody binding assays, assay development, and QC testing.
- Size: 25 μg.
- Format: Lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
- Expression system: HEK293 (human) cells.
- Tags: C-terminal His tag and Avi tag.
- Conjugate: Biotinylated via Avitag technology.
- Purity: >90% by SDS-PAGE and SEC-MALS.
- Molecular weight: calculated 36.0 kDa (HLA) and 11.7 kDa (B2M); observed ~40-45 kDa and ~12 kDa by SDS-PAGE.
- Storage: Lyophilized at -20°C or lower (12 months); after reconstitution store at -70°C (up to 3 months).
- Applications: Validated for binding to anti-HLA class I and anti-B2M antibodies (linear range 0.1-2 ng/mL).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.