Learn More
ACROBiosystems Biotinylated human HLA-A*11:01 & B2M complex with KRAS G12V peptide (VVGAVGVGK), 200 μg, AviTag/His
Supplier:  ACROBiosystems HLVH82E6200UG
Encompass_Preferred
Biotinylated human HLA-A*11:01 complexed with β2-microglobulin and the KRAS G12V peptide (VVGAVGVGK). Monomeric complex produced in HEK293 cells and enzymatically biotinylated at an Avi tag; supplied lyophilized for use in antigen presentation, T-cell receptor binding, and immune assay development.
  • Monomeric HLA-A*11:01-B2M complex loaded with KRAS G12V peptide (VVGAVGVGK).
  • Enzymatically biotinylated at the Avi tag for site-specific labeling.
  • C-terminal polyhistidine tag for purification.
  • Lyophilized from PBS with trehalose; reconstitute in PBS, pH 7.4.
  • Purity >90% by SDS-PAGE and >90% by SEC-MALS.
  • 200 μg total (100 μg x 2); stable 12 months lyophilized at -20°C to -70°C.
  • Validated by SEC-MALS; suitable for binding and immunological assays.
Catalog No. 50-237-0239