Learn More
ACROBiosystems Biotinylated Human HLA-A*11:01 & B2M & KRAS G12D (VVGADGVGK) Complex Protein, site-specific biotinylated, 200 μg (Monomer, MALS verified)
Catalog No. 502370247
Encompass_Preferred
Biotinylated Human HLA-A*11:01 & B2M & KRAS G12D (VVGADGVGK) complex protein is a monomeric MHC class I complex, co-expressed and loaded with the KRAS G12D peptide. It is site-specifically enzymatically biotinylated, supplied lyophilized (200 μg), and validated for binding assays including ELISA and SPR.
  • Monomeric MHC class I complex comprising HLA-A*11:01, beta-2 microglobulin, and KRAS G12D peptide.
  • Expressed in human HEK293 cells to preserve mammalian glycosylation.
  • Site-specific enzymatic biotinylation for consistent capture on streptavidin surfaces.
  • Supplied lyophilized (200 μg) formulated in PBS with trehalose for stability.
  • Purity greater than 90% by SDS-PAGE and by SEC-MALS; monomeric state verified by MALS.
  • Validated in ELISA and SPR; SPR affinity reported ~1.13 nM in routine testing.
  • Stable at -20°C to -70°C in lyophilized form; reconstituted material stable at -70°C for short-term storage.
Catalog No. 50-237-0247 Supplier ACROBiosystems Supplier No. HLDH82E5200UG
Biotinylated Human HLA-A*11:01 & B2M & KRAS G12D (VVGADGVGK) complex protein is a monomeric MHC class I complex, co-expressed and loaded with the KRAS G12D peptide. It is site-specifically enzymatically biotinylated, supplied lyophilized (200 μg), and validated for binding assays including ELISA and SPR.
  • Monomeric MHC class I complex comprising HLA-A*11:01, beta-2 microglobulin, and KRAS G12D peptide.
  • Expressed in human HEK293 cells to preserve mammalian glycosylation.
  • Site-specific enzymatic biotinylation for consistent capture on streptavidin surfaces.
  • Supplied lyophilized (200 μg) formulated in PBS with trehalose for stability.
  • Purity greater than 90% by SDS-PAGE and by SEC-MALS; monomeric state verified by MALS.
  • Validated in ELISA and SPR; SPR affinity reported ~1.13 nM in routine testing.
  • Stable at -20°C to -70°C in lyophilized form; reconstituted material stable at -70°C for short-term storage.