Learn More
Invitrogen™ Protein A
Description
Protein A purified from Staphylococcus aureus. Lyophilized, salt-free powder; no preservative added. Molecular weight is approximately 42kD.
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C. |
| Shipping Condition | Wet Ice |
| Label or Dye | Unconjugated |
| Type | Protein A |
| Quantity | 25 MG |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.