Learn More
Seracare Life Sciences Inc PROTEIN A AGAROSE
Supplier:  Seracare Life Sciences Inc 57100005
Encompass_Preferred
Protein A Agarose consists of native protein A immobilized onto 4% cross linked agarose beads. It is designed specifically for the binding of immunoglobulins for both laboratory and process scale applications. Protein A binds specifically to the Fc region of immunoglobulin molecules of many mammalian species without disturbing their binding of antigen.
Catalog No. 50-673-76