Learn More
Gold Biotechnology Inc Protein A Agarose Resin
SDP
Supplier:  Gold Biotechnology Inc P4005
Encompass_Preferred
Protein A agarose resin consists of rProtein A covalently bound to cross-linked agarose beads It provides a very stable bond that can greatly minimize leakage of the Protein A allowing for reuse of the affinity resin in several purification steps Protein A Agarose resin is available in bulk from 5-100 ml quantities
Catalog No. 50-005-62069