Learn More
Gold Biotechnology Inc Protein A Agarose Resin
SDP
Catalog No. 5000562104
Encompass_Preferred
Protein A agarose resin consists of rProtein A covalently bound to cross-linked agarose beads It provides a very stable bond that can greatly minimize leakage of the Protein A allowing for reuse of the affinity resin in several purification steps Protein A Agarose resin is available in bulk from 5-100 ml quantities
Catalog No. 50-005-62104 Supplier Gold Biotechnology Inc Supplier No. P40050
Add to Cart
Add to Cart
Protein A agarose resin consists of rProtein A covalently bound to cross-linked agarose beads It provides a very stable bond that can greatly minimize leakage of the Protein A allowing for reuse of the affinity resin in several purification steps Protein A Agarose resin is available in bulk from 5-100 ml quantities