Learn More
Cygnus Technologies Cygnus Technologies Protein A Antigen Concentrate, Positive Control for ELISA, Spike and Recovery Experiments, 26 units
Supplier:  Cygnus Technologies F613X
Encompass
Cygnus Technologies Protein A Antigen Concentrate is designed for scientific applications requiring precise detection and quantification of Protein A. It serves as a crucial component in spike and recovery experiments and as a positive control for the Protein A Mix-N-Go ELISA Kit. This product enhances the accuracy of assessing residual Protein A levels in biopharmaceutical processes.
Catalog No. NC2035481