Learn More
Bioworld PROTEIN-A BLUE SEPAROPORE
Catalog No. 50197772
Encompass
PROTEIN-A BLUE SEPAROPORE (AGAROSE) 4B (MACROBEADS): Protein A Blue Separopore 4B (Macrobeads) Affinity matrix with immobilized Protein-A by CNBr activation coupling method.; For purification and/or fractionation of IgG subclasses.; Rel...
Catalog No. 50-197-772 Supplier Bioworld Supplier No. 201810423
PROTEIN-A BLUE SEPAROPORE (AGAROSE) 4B (MACROBEADS): Protein A Blue Separopore 4B (Macrobeads) Affinity matrix with immobilized Protein-A by CNBr activation coupling method.; For purification and/or fractionation of IgG subclasses.; Rel...