Learn More
Bioworld Protein A-Coated Microplates, 5 Plate(s)
Catalog No. 50197543
Encompass
Protein A coatedmicroplates provide alternative methods for immobilizing antibodies for ELISA and other plate-based assay techniques. These plates are uniformly and stably coated with Protein A. They enable antibodies or pre-estabilished antibody-antigen complexes to be captured to wells of the polystyrene plate for subsequent detection in plate readers. The strategy is useful for a variety of assay situations, but it is generally not suitable for sandwich-type ELISA.Specifications:Retain antibody activity, which can be lost when antibodies are immobilized by passive adsorption Orient antibodies for maximum antigen-binding capacity Immobilize antibodies without prior purification Ensure minimal variation (<5% well-to-well) from consistent coating Reduce nonspecific binding because plates are pre-blockedBinds up to 2.5µg antibody/well (100µL coating volume)
Catalog No. 50-197-543 Supplier Bioworld Supplier No. 201400121
Protein A coatedmicroplates provide alternative methods for immobilizing antibodies for ELISA and other plate-based assay techniques. These plates are uniformly and stably coated with Protein A. They enable antibodies or pre-estabilished antibody-antigen complexes to be captured to wells of the polystyrene plate for subsequent detection in plate readers. The strategy is useful for a variety of assay situations, but it is generally not suitable for sandwich-type ELISA.Specifications:Retain antibody activity, which can be lost when antibodies are immobilized by passive adsorption Orient antibodies for maximum antigen-binding capacity Immobilize antibodies without prior purification Ensure minimal variation (<5% well-to-well) from consistent coating Reduce nonspecific binding because plates are pre-blockedBinds up to 2.5µg antibody/well (100µL coating volume)