Learn More
Enzo Life Sciences Protein A ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900057
Encompass_Preferred
Most sensitive (9.01 pg/ml)  colorimetric ELISA for monitoring residual contamination. The Protein A EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Ultra-sensitive quantification ( < 1 ppm ) of Protein A residuals in purified humanized monoclonal antibody preparations. This kit recognizes four different Protein A constructs. Sensitivity: 9.01 pg/ml (range 15.63 - 1,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate,Standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-120