Learn More
Genscript Corporation Antibody Kit Protein Analysis Protein Analysis 1/EA
Supplier:  Genscript Corporation L00430
Encompass
Protein A affinity chromatography is widely used for antibody purification. However the leakage of Protein A though covalently conjugated to chromatography supports posts a big problem for antibody based pharmaceutical industry where false results could be generated in immunological assays from contaminated Protein A.
Catalog No. 50-252-9997